Learn More
Chemglass Life Sciences Pipette Filler, Manual, Quick-Release, for up to 10mL, Green
SDP
Catalog No. 501897597
Encompass
Designed for quick filling and releasing of liquid in aspirating and serological pipettes. Easy to use and easy to clean. Simply rotate the thumb wheel to aspirate or dispense accurate measurements. Press the release lever to quickly dispense all liquid from the pipettes. Resistant to acids and alkalies.
Catalog No. 50-189-7597 Supplier Chemglass Life Sciences Supplier No. CGE347251
Designed for quick filling and releasing of liquid in aspirating and serological pipettes. Easy to use and easy to clean. Simply rotate the thumb wheel to aspirate or dispense accurate measurements. Press the release lever to quickly dispense all liquid from the pipettes. Resistant to acids and alkalies.