Learn More
Cole-Parmer Argos Technologies Manual Pipette Pump, 25 mL, Red
Catalog No. 501960557 Shop All Cole-Parmer Products
Encompass_Preferred
These manual pipette pumps feature a thumbwheel for precision filling or dispensing of liquids. Insert glass or plastic pipettes into the secure tapered collar andthen quickly dispense liquid by depressing its white side bar or plunger. The pumps can operate easily with one hand.
Catalog No. 50-196-0557 Supplier Cole-Parmer Supplier No. 0439506
These manual pipette pumps feature a thumbwheel for precision filling or dispensing of liquids. Insert glass or plastic pipettes into the secure tapered collar andthen quickly dispense liquid by depressing its white side bar or plunger. The pumps can operate easily with one hand.