Learn More
Chemglass Life Sciences Pipette Filler, Manual, Quick-Release, for up to 2mL, Blue
SDP
Catalog No. 501897596
Encompass
Designed for quick filling and releasing of liquid in aspirating and serological pipettes. Easy to use and easy to clean. Simply rotate the thumb wheel to aspirate or dispense accurate measurements. Press the release lever to quickly dispense all liquid from the pipettes. Resistant to acids and alkalies.
Catalog No. 50-189-7596 Supplier Chemglass Life Sciences Supplier No. CGE347250
Designed for quick filling and releasing of liquid in aspirating and serological pipettes. Easy to use and easy to clean. Simply rotate the thumb wheel to aspirate or dispense accurate measurements. Press the release lever to quickly dispense all liquid from the pipettes. Resistant to acids and alkalies.