Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fischer Technical Company™ Pipette Filler Assortment Set of 4
SCI_ED

Catalog No. S2335028 Shop All Fischer Technical Company Products
Change view
Click to view available options
Volume (Metric):
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
S2335028 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S2335028 Supplier Fischer Technical Company™ Supplier No. PFKIT4
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Convenient assortment of four different types of pipette fillers.

Includes one natural rubber red three valve pipette filler, one silicon three valve pipette filler, one red natural rubber two valve Flip style pipette filler, and one natural rubber three valve multicolored pipette filler.

  • Accommodates pipettes up to 100 mL (bulbs draw 50 mL at a time, for volumes larger than 50 mL user will have to repeat the liquid draw process)

Specifications

Adjustable Tip Spacing No
Autoclavable Not Autoclavable
Color Assorted
Disposable Reusable
For Use With Pipette
Grip Type Bulb
Material Natural Rubber
Model PFKIT4
Number of Channels 1
Sterility Non-sterile
Type Pipette Filler
Volume (Metric) 100 mL
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.