Learn More
Research Products International Corp Pipette Filler, Mini
SDP
Catalog No. 501978114
Encompass
RPI Mini-Pipette Filler offers lightweight aspiration and dispensing with 0.1 ml up to 100 ml size pipettes. Allows precise pipette control during operation. Mini squeeze bulb is provided for blow out style pipettors. Supplied with 3 m filter.
Catalog No. 50-197-8114 Supplier Research Products International Corp Supplier No. 248649
RPI Mini-Pipette Filler offers lightweight aspiration and dispensing with 0.1 ml up to 100 ml size pipettes. Allows precise pipette control during operation. Mini squeeze bulb is provided for blow out style pipettors. Supplied with 3 m filter.