Learn More
Chemglass Life Sciences Motorized Pipette Filler
SDP
Catalog No. 50958489
Encompass_Preferred
Motorized Pipette Filler. The motorized Pipette Filler features a full volume range of 0.1 to 100mL with easy one-handed operation. Its lightweight, ergonomic design allows longer, fatigue-free pipetting with separate aspirate and dispense speed controls. A large LCD display provides visual confirmation of remaining battery charge and speed settings. The lithium-ion battery offers long runtimes per charge without battery memory problems. The fully autoclavable nosecone housing simplifies disinfecting and decontamination.
Catalog No. 50-958-489 Supplier Chemglass Life Sciences Supplier No. CLS347210
Motorized Pipette Filler. The motorized Pipette Filler features a full volume range of 0.1 to 100mL with easy one-handed operation. Its lightweight, ergonomic design allows longer, fatigue-free pipetting with separate aspirate and dispense speed controls. A large LCD display provides visual confirmation of remaining battery charge and speed settings. The lithium-ion battery offers long runtimes per charge without battery memory problems. The fully autoclavable nosecone housing simplifies disinfecting and decontamination.