Learn More
Electron Microscopy Sciences Quick-Release Pipette Filler 10 ml, Green
SDP
Catalog No. 501899383
Encompass_Preferred
This pipetting device pro-vides an easy, precise way of filling and releasing liquid. By rotating the knurled thumb wheel the liquid is drawn up or dispensed. The soft elastic chuck has a threaded collar that secures the plastic or glass pipette to the device. Sizes are color coded. Fits standard or disposable pipettes and diposable serological tooled-mouthpiece pipettes as per ASTM Standard E714-80.
Color: Green
Catalog No. 50-189-9383 Supplier Electron Microscopy Sciences Supplier No. 70958
This pipetting device pro-vides an easy, precise way of filling and releasing liquid. By rotating the knurled thumb wheel the liquid is drawn up or dispensed. The soft elastic chuck has a threaded collar that secures the plastic or glass pipette to the device. Sizes are color coded. Fits standard or disposable pipettes and diposable serological tooled-mouthpiece pipettes as per ASTM Standard E714-80.
Color: Green