Learn More
Sigma Aldrich Fine Chemicals Biosciences Pipette fillers red natura1EA
Catalog No. 502900939
Encompass
Valved choice of natural or silicone rubber bulb. Allows safe manual control of filling and discharging of liquids by finger pressure.
Catalog No. 50-290-0939 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z1364681EA
Valved choice of natural or silicone rubber bulb. Allows safe manual control of filling and discharging of liquids by finger pressure.