Learn More
Electron Microscopy Sciences Pipette Filter Tips, 5-10 mcL, Sterile, Natural color
SDP
Catalog No. 501899939
Encompass_Preferred
Kendall filter pipette tips are certified Rnase / Dnaee-Free, human DNA-Free, and PCR Inhibitor-Free. The low binding surface reduces DNA and protection retention. Liquid and aerosol barrier prevent pipetter contamination. A universal fit on most research-grade pipetters. CellPack keeps tips straight for multi-channel pipetters. Filter made of high-density polyethylene (naturally hydrophobic). Types of membranes are an aerosol barrier to prevents aerosol contamination, a self-sealing Barrier (contains cellulose) that prevents aerosol and liquid contamination and cellulose can contaminate samples, and a liquid barrier that prevents aerosol and liquid contamination and will not contaminate samples.
Range: 5-10 µl
Sterile: Sterile
Catalog No. 50-189-9939 Supplier Electron Microscopy Sciences Supplier No. 7246003
Kendall filter pipette tips are certified Rnase / Dnaee-Free, human DNA-Free, and PCR Inhibitor-Free. The low binding surface reduces DNA and protection retention. Liquid and aerosol barrier prevent pipetter contamination. A universal fit on most research-grade pipetters. CellPack keeps tips straight for multi-channel pipetters. Filter made of high-density polyethylene (naturally hydrophobic). Types of membranes are an aerosol barrier to prevents aerosol contamination, a self-sealing Barrier (contains cellulose) that prevents aerosol and liquid contamination and cellulose can contaminate samples, and a liquid barrier that prevents aerosol and liquid contamination and will not contaminate samples.
Range: 5-10 µl
Sterile: Sterile