Learn More
Narishige International Usa Inc PIPETTE HOLDER 4MM BAR HOLDER
Catalog No. NC2007722
Encompass
H-7 Pipette Holder ø4mm bar holder for ø1 ~ 1.5mm pipettes.
Catalog No. NC2007722 Supplier Narishige International Usa Inc Supplier No. H7
H-7 Pipette Holder ø4mm bar holder for ø1 ~ 1.5mm pipettes.