Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Electron Microscopy Sciences Narrow Stem Pipettes 8.7 ml, 4.8 ml Bulb draw, Sterile individual package
SDP

Catalog No. 501899412
Encompass_Preferred

These polyethylene disposable have a low-affinity surface, attached bulb avoids cross-contamination, and are one step biohazard disposable. These Micro-tip pipets are ideal for loading gels, DNA sample removal, protein assays, lipids extractions, drop-wise additions to culture plates, using 2 ml auto sampler vials, remove or disperse small volumes etc.
Bulb Draw: 4.8 ml
Drops/mL: 45
OAL: 14.7 cm
Stem diameter: 6.1 cm
Package: Individual

Catalog No. 50-189-9412 Supplier Electron Microscopy Sciences Supplier No. 7096713S
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

These polyethylene disposable have a low-affinity surface, attached bulb avoids cross-contamination, and are one step biohazard disposable. These Micro-tip pipets are ideal for loading gels, DNA sample removal, protein assays, lipids extractions, drop-wise additions to culture plates, using 2 ml auto sampler vials, remove or disperse small volumes etc.
Bulb Draw: 4.8 ml
Drops/mL: 45
OAL: 14.7 cm
Stem diameter: 6.1 cm
Package: Individual

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.