Learn More
Research Products International Corp Fast-Release Pipette Pump, Green, Fits Pipettes up to 10 ml
SDP
Catalog No. 50212732
Encompass
Pipette Pump provides easy and accurate one-hand pipetting. Specially designed to offer precise alignment, secure grip and locking of glass or plastic pipettes. Features rapid release lever for quick emptying and thumbwheel for drawing and dispensing liquid.
Catalog No. 50-212-732 Supplier Research Products International Corp Supplier No. 437910
Pipette Pump provides easy and accurate one-hand pipetting. Specially designed to offer precise alignment, secure grip and locking of glass or plastic pipettes. Features rapid release lever for quick emptying and thumbwheel for drawing and dispensing liquid.