Learn More
Research Products International Corp Fast-Release Pipette Pump, Red, Fits Pipettes Up to 25 ml
SDP
Catalog No. 50212733
Encompass
Pipette Pump provides easy and accurate one-hand pipetting. Specially designed to offer precise alignment, secure grip and locking of glass or plastic pipettes. Features rapid release lever for quick emptying and thumbwheel for drawing and dispensing liquid.
Catalog No. 50-212-733 Supplier Research Products International Corp Supplier No. 437925
Pipette Pump provides easy and accurate one-hand pipetting. Specially designed to offer precise alignment, secure grip and locking of glass or plastic pipettes. Features rapid release lever for quick emptying and thumbwheel for drawing and dispensing liquid.