Learn More
Sigma Aldrich Fine Chemicals Biosciences Pipette PumpTM II blue1EA
Catalog No. 502901730
Encompass
for safe and accurate pipetting with one-handed operation. Rotation of the thumb wheel provides precise control of filling or dispensing. Rapid emptying is possible by pressing the plunger or the quick release bar (not available on the 0.2AmL pump). Color coded for easy identification.
Catalog No. 50-290-1730 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. P80491EA
for safe and accurate pipetting with one-handed operation. Rotation of the thumb wheel provides precise control of filling or dispensing. Rapid emptying is possible by pressing the plunger or the quick release bar (not available on the 0.2AmL pump). Color coded for easy identification.