Learn More
Electron Microscopy Sciences SureStand™ Pipette Rack 6 Slots with 4 Slots for Multi-Channel
SDP
Catalog No. 501931083
Encompass_Preferred
SureStand Pipette racks are unique in their ability to accommodate multi-channel pipettes without sacrificing any of the stability of a traditional acrylic rack. The patent pending "windows" allow the user to insert the multi-channel head longways through the rack, while keeping the body of the pipette stable by securing the pipette at two points; under the finger hook, and near the bottom of the handle. Each SureStand is also outfitted with four non-skid rubber "feet" for extra stability. SureStand is guaranteed to be the sturdiest and most versatile pipette rack in your lab. 6-place rack that accepts four multi-channel pipette slots.
Catalog No. 50-193-1083 Supplier Electron Microscopy Sciences Supplier No. 6436303
SureStand Pipette racks are unique in their ability to accommodate multi-channel pipettes without sacrificing any of the stability of a traditional acrylic rack. The patent pending "windows" allow the user to insert the multi-channel head longways through the rack, while keeping the body of the pipette stable by securing the pipette at two points; under the finger hook, and near the bottom of the handle. Each SureStand is also outfitted with four non-skid rubber "feet" for extra stability. SureStand is guaranteed to be the sturdiest and most versatile pipette rack in your lab. 6-place rack that accepts four multi-channel pipette slots.