Learn More
Med Vet International McKesson Serological Pipette, 10 mL, 177-1761, 200/Case
Supplier:  Med Vet International 1176007CS
Encompass_Preferred
McKesson Serological Pipette 10 mL 0.1 mL Graduation Increments / 2.5 mL Negative Graduations Sterile Sterile Standard tip Orange striped 10 mL with 0.1 mL graduation increments Calibration is certified for accuracy Aerosol barrier plug Reverse graduations and negative graduations Fits major brands of pipette controllers Designed with anti-drip tips Color coding makes it easy to identify the size needed Polystyrene
Catalog No. 50-304-4100