Learn More
Med Vet International McKesson Polystyrene Serological Pipette, 50 mL, 177-1821, 100/Case
Catalog No. 503044937
Encompass_Preferred
McKesson Serological Pipette 50 mL 0.5 mL Graduation Increments / 9 mL Negative Graduations Sterile 0.5 mL graduation increments / 9 mL negative graduations Calibration is certified for accuracy Aerosol barrier plug Fits major brands of pipette controllers Designed with anti-drip tips Standard tip, black striped Color coding makes it easy to identify the size needed Made of polystyrene There are 100 individually packaged, 50 mL serological pipettes per case
Catalog No. 50-304-4937 Supplier Med Vet International Supplier No. 1176009CS
McKesson Serological Pipette 50 mL 0.5 mL Graduation Increments / 9 mL Negative Graduations Sterile 0.5 mL graduation increments / 9 mL negative graduations Calibration is certified for accuracy Aerosol barrier plug Fits major brands of pipette controllers Designed with anti-drip tips Standard tip, black striped Color coding makes it easy to identify the size needed Made of polystyrene There are 100 individually packaged, 50 mL serological pipettes per case