Learn More
Pipette Supplies, Inc. Gilson original piston assembly 1000uL
SDP
Catalog No. NC1844272
Encompass
Gilson piston assembly for: Gilson Pipetman Classic 1000μL (P1000), Gilson Pipetman Fixed F250, F300, F400, F500, F1000, Gilson Pipetman Neo 1000μL (P1000N)
Catalog No. NC1844272 Supplier Pipette Supplies, Inc. Supplier No. GI F123847
Gilson piston assembly for: Gilson Pipetman Classic 1000μL (P1000), Gilson Pipetman Fixed F250, F300, F400, F500, F1000, Gilson Pipetman Neo 1000μL (P1000N)