Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Pipette Stand
SCI_ED

Catalog No. S29071 Shop All Fisherbrand Products
Change view
Click to view available options
For Use With (Equipment):
Multichannel Pipette
Single-Channel Pipette
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
S29071 Multichannel Pipette
S29069 Single-Channel Pipette
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. S29071
This item is not returnable. View return policy

Accommodate most major brands of pipetters.

  • One-position stand has a single insert hole 1.25 in. (31.8 mm) in diameter
  • Three-, four-, and six-position stands have both upper (1.03 in. [26.1 mm]) and lower slots (0.625 in. [16 mm] in diameter)
  • Multichannel stand is clear acrylic with slots 1 in. (25.5 mm) in diameter

Specifications

Autoclavable Not Autoclavable
Color Clear
Holds Multichannel Pipettor
Material Acrylic
Positions 1
For Use With (Equipment) Multichannel Pipette
Dimensions (L x W x H) 14.2 x 7.6 x 24.1 cm (5.6 x 3 x 9.5 in.)
Product Type Pipet Stand
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.