Learn More
Chemglass Life Sciences Pipette Storage Rack, Magnetized, White
SDP
Catalog No. 501532315
Encompass_Preferred
Pipette Storage Rack, Magnetized, White. This magnetized pipette storage rack has strong magnets on both sides and mounts securely on fume hoods, file cabinets, refrigerators, etc. Constructed of durable ABS plastic, this rack not only looks better, but will also hold up better than acrylic. Includes two handy attachments for holding pipette bulbs, pipette fillers and pipette pumps. Measures 290 x 85 x 395mm.
Catalog No. 50-153-2315 Supplier Chemglass Life Sciences Supplier No. CLS347601
Pipette Storage Rack, Magnetized, White. This magnetized pipette storage rack has strong magnets on both sides and mounts securely on fume hoods, file cabinets, refrigerators, etc. Constructed of durable ABS plastic, this rack not only looks better, but will also hold up better than acrylic. Includes two handy attachments for holding pipette bulbs, pipette fillers and pipette pumps. Measures 290 x 85 x 395mm.