Learn More
Electron Microscopy Sciences Pipette Tip P2/P10 Ultra Micro, 0.1 -10 mcL, Sterile
SDP
Catalog No. 501899904
Encompass_Preferred
Universal fit for ultra-micro pipetters with graduation marks at 2 ul and 5 ul. Without low binding surface.
Range: 0.1 -10 µl
Sterile: Sterile
Catalog No. 50-189-9904 Supplier Electron Microscopy Sciences Supplier No. 7244004
Universal fit for ultra-micro pipetters with graduation marks at 2 ul and 5 ul. Without low binding surface.
Range: 0.1 -10 µl
Sterile: Sterile