Learn More
Electron Microscopy Sciences Pipette Tip Universal, 101-1000 mcL, Sterile
SDP
Catalog No. 501899921
Encompass_Preferred
Pipette tips are low binding to ensure accuracy and precision. The low binding surface is the result of a manufacturing process that binds a perfluorooctyl polymer to polypropylene resin–resulting in an exposed trifluoromethyl surface. This surface reduces the surface energy of liquid and repels any substance so sample retention and binding are minimized.
Not detectable/Non-reactive.
Extracted only by a fluorinated solvent.
Autoclaving will not compromise performance.
Available in convenient hinged racked package.
Tips and racks are fully autoclavable. Natural tips feature reference marks at 10 ul, 50 ul, and 100 u. Low binding surface.
Range: 101-1000 µl
Sterile: Sterile
Catalog No. 50-189-9921 Supplier Electron Microscopy Sciences Supplier No. 7244503
Pipette tips are low binding to ensure accuracy and precision. The low binding surface is the result of a manufacturing process that binds a perfluorooctyl polymer to polypropylene resin–resulting in an exposed trifluoromethyl surface. This surface reduces the surface energy of liquid and repels any substance so sample retention and binding are minimized.
Not detectable/Non-reactive.
Extracted only by a fluorinated solvent.
Autoclaving will not compromise performance.
Available in convenient hinged racked package.
Tips and racks are fully autoclavable. Natural tips feature reference marks at 10 ul, 50 ul, and 100 u. Low binding surface.
Range: 101-1000 µl
Sterile: Sterile