Learn More
Electron Microscopy Sciences Pipette Tip Universal, 101-1000 mcL
SDP
Catalog No. 501899920
Encompass_Preferred
Pipette tips are low binding to ensure accuracy and precision. The low binding surface is the result of a manufacturing process that binds a perfluorooctyl polymer to polypropylene resin–resulting in an exposed trifluoromethyl surface. This surface reduces the surface energy of liquid and repels any substance so sample retention and binding are minimized.
Not detectable/Non-reactive.
Extracted only by a fluorinated solvent.
Autoclaving will not compromise performance.
Available in convenient hinged racked package.
Tips and racks are fully autoclavable. Natural tips feature reference marks at 10 ul, 50 ul, and 100 u. Low binding surface.
Range: 101-1000 µl
Catalog No. 50-189-9920 Supplier Electron Microscopy Sciences Supplier No. 7244502
Pipette tips are low binding to ensure accuracy and precision. The low binding surface is the result of a manufacturing process that binds a perfluorooctyl polymer to polypropylene resin–resulting in an exposed trifluoromethyl surface. This surface reduces the surface energy of liquid and repels any substance so sample retention and binding are minimized.
Not detectable/Non-reactive.
Extracted only by a fluorinated solvent.
Autoclaving will not compromise performance.
Available in convenient hinged racked package.
Tips and racks are fully autoclavable. Natural tips feature reference marks at 10 ul, 50 ul, and 100 u. Low binding surface.
Range: 101-1000 µl