Learn More
Chemglass Life Sciences Pipette Tip, 10µL, Extra Long, Barrier Tip, Sterile, 960/Case
SDP
Catalog No. 501532325
Encompass_Preferred
Pipette Tip, 10¿L, Extra Long, Barrier Tip, Sterile, 960/Case. Filtered pipette tips are the experienced scientist¿s choice for eliminating carryover contamination in the laboratory environment. Aerosol barrier filters are molded with ultra-high molecular weight polyethylene, completely inert and consistent in porosity. This ensures that the filter/barrier within the tips will not impede airflow and will protect your samples and pipettor from harmful contamination. The filters are manufactured within a clean-room facility, therefore every component is free of nucleic acid and enzymes which are detrimental to your research. All the filtered tips are pre-sterilized and certified by an independent lab to be free of nucleic acids, RNase, DNase and pyrogens.
Catalog No. 50-153-2325 Supplier Chemglass Life Sciences Supplier No. CLS3499C010
Pipette Tip, 10¿L, Extra Long, Barrier Tip, Sterile, 960/Case. Filtered pipette tips are the experienced scientist¿s choice for eliminating carryover contamination in the laboratory environment. Aerosol barrier filters are molded with ultra-high molecular weight polyethylene, completely inert and consistent in porosity. This ensures that the filter/barrier within the tips will not impede airflow and will protect your samples and pipettor from harmful contamination. The filters are manufactured within a clean-room facility, therefore every component is free of nucleic acid and enzymes which are detrimental to your research. All the filtered tips are pre-sterilized and certified by an independent lab to be free of nucleic acids, RNase, DNase and pyrogens.