Learn More
Chemglass Life Sciences Chemglass Pipette Tip, 10 μL Extra-Long Filtered (Sterile) - Bulk 4,800/Case; for single and multichannel pipettors
SDP
Catalog No. 502733219
Encompass_Preferred
Chemglass extra-long 10 μL filtered pipette tips are manufactured from premium 100% polypropylene for precise, low-volume liquid handling in molecular biology and analytical laboratories. Gamma-irradiated sterile and certified free of RNase, DNase, human DNA, and pyrogens, these fine-point barrier tips reduce sample carryover and protect pipettors from contamination.
  • Extra-long, fine-point design for precise low-volume dispensing.
  • Hydrophobic aerosol barrier reduces carryover and contamination risk.
  • Gamma-irradiated sterile and certified RNase-, DNase-, human DNA-, and pyrogen-free.
  • Made from 100% polypropylene with low-retention surface for minimal sample adhesion.
  • Graduated for quick volume checks.
  • Universal-fit for most single and multichannel pipettors.
  • Bulk packed, 4,800 tips per case.
Catalog No. 50-273-3219 Supplier Chemglass Life Sciences Supplier No. CLS3499D010
Chemglass extra-long 10 μL filtered pipette tips are manufactured from premium 100% polypropylene for precise, low-volume liquid handling in molecular biology and analytical laboratories. Gamma-irradiated sterile and certified free of RNase, DNase, human DNA, and pyrogens, these fine-point barrier tips reduce sample carryover and protect pipettors from contamination.
  • Extra-long, fine-point design for precise low-volume dispensing.
  • Hydrophobic aerosol barrier reduces carryover and contamination risk.
  • Gamma-irradiated sterile and certified RNase-, DNase-, human DNA-, and pyrogen-free.
  • Made from 100% polypropylene with low-retention surface for minimal sample adhesion.
  • Graduated for quick volume checks.
  • Universal-fit for most single and multichannel pipettors.
  • Bulk packed, 4,800 tips per case.