Learn More
Chemglass Life Sciences Chemglass 10 μL Sterile Barrier Pipette Tip, Standard - Bulk 4,800/Case
SDP
Catalog No. 502733218
Encompass_Preferred
Sterile 10 μL barrier pipette tips designed for accurate micro-volume dispensing and to protect pipettors from contamination. Gamma-irradiated and certified RNase-, DNase-, human DNA-, and pyrogen-free, these ultra-clear polypropylene tips provide low-retention performance and universal-fit compatibility.
  • Sterile, gamma-irradiated and certified free of RNase, DNase, human DNA, and pyrogens.
  • 10 μL nominal volume for micro-volume pipetting.
  • Standard tip profile with fine-point design for low sample adhesion.
  • Hydrophobic aerosol barrier to prevent carry-over and contamination.
  • Ultra-clear, premium-grade polypropylene construction.
  • Bulk packed, 4,800 tips per case for high-volume use.
  • Universal-fit for most single and multichannel pipettors.
Catalog No. 50-273-3218 Supplier Chemglass Life Sciences Supplier No. CLS3499D018
Add to Cart
Add to Cart
Sterile 10 μL barrier pipette tips designed for accurate micro-volume dispensing and to protect pipettors from contamination. Gamma-irradiated and certified RNase-, DNase-, human DNA-, and pyrogen-free, these ultra-clear polypropylene tips provide low-retention performance and universal-fit compatibility.
  • Sterile, gamma-irradiated and certified free of RNase, DNase, human DNA, and pyrogens.
  • 10 μL nominal volume for micro-volume pipetting.
  • Standard tip profile with fine-point design for low sample adhesion.
  • Hydrophobic aerosol barrier to prevent carry-over and contamination.
  • Ultra-clear, premium-grade polypropylene construction.
  • Bulk packed, 4,800 tips per case for high-volume use.
  • Universal-fit for most single and multichannel pipettors.