Learn More
Chemglass Life Sciences Chemglass Pipette Tip, 1250 μL, Standard barrier, Sterile, universal-fit - 960/Case
SDP
Catalog No. 502733221
Encompass_Preferred
Sterile 1250 μL barrier pipette tips designed for accurate, low-retention liquid handling in molecular biology and general laboratory workflows. Gamma-irradiated and certified free of RNase, DNase, human DNA, and pyrogens; packed 960 tips per case for high-throughput use.
  • Tip volume: 1250 μL
  • Material: 100% polypropylene
  • Sterility: Sterile, gamma-irradiated
  • Certifications: RNase/DNase-free, human DNA-free, pyrogen-free
  • Pack quantity: 960 per case
  • Design features: Low-retention, fine-point, graduated
  • Barrier: Hydrophobic aerosol barrier to reduce carry-over
  • Compatibility: Universal-fit for most single- and multichannel pipettors
Catalog No. 50-273-3221 Supplier Chemglass Life Sciences Supplier No. CLS3499C1250
Sterile 1250 μL barrier pipette tips designed for accurate, low-retention liquid handling in molecular biology and general laboratory workflows. Gamma-irradiated and certified free of RNase, DNase, human DNA, and pyrogens; packed 960 tips per case for high-throughput use.
  • Tip volume: 1250 μL
  • Material: 100% polypropylene
  • Sterility: Sterile, gamma-irradiated
  • Certifications: RNase/DNase-free, human DNA-free, pyrogen-free
  • Pack quantity: 960 per case
  • Design features: Low-retention, fine-point, graduated
  • Barrier: Hydrophobic aerosol barrier to reduce carry-over
  • Compatibility: Universal-fit for most single- and multichannel pipettors