Learn More
Chemglass Life Sciences Chemglass pipette tip 1,250 μL - sterile aerosol-barrier tip for large-volume pipettors (CLS-3499D-1250)
SDP
Catalog No. 502733216
Encompass_Preferred
Sterile 1,250 μL pipette tips with integrated aerosol barrier filters provide contamination protection for large-volume pipetting and reliable liquid handling in laboratory workflows.
  • Sterile.
  • Integrated aerosol barrier filter.
  • 1,250 μL capacity.
  • Standard fit for large-volume pipettors.
  • Helps reduce aerosol formation and cross-contamination during transfer.
Catalog No. 50-273-3216 Supplier Chemglass Life Sciences Supplier No. CLS3499D1250
Sterile 1,250 μL pipette tips with integrated aerosol barrier filters provide contamination protection for large-volume pipetting and reliable liquid handling in laboratory workflows.
  • Sterile.
  • Integrated aerosol barrier filter.
  • 1,250 μL capacity.
  • Standard fit for large-volume pipettors.
  • Helps reduce aerosol formation and cross-contamination during transfer.