Learn More
Cole-Parmer Essentials Pipette Tip, 20 to 200 µL, Bulk; 1000/BG
Catalog No. NC1971806 Shop All Cole-Parmer Products
Encompass_Preferred
Not your everyday graduated pipette tips. It starts with a touch-free manufacturing facility and stringent quality control process to make these clear, plastic tips certified free of any affecting biological elements, such as human DNA, DNase and RNase and Endotoxin, or Pyrogen and PCR inhibitors. They also are noncytotoxic. It’s about their innovative design, too. The profile and dimensions of the tips are engineered to accurately measure and transfer a volume of liquid from one container to another, as well as adapt to a wide-range of pipettes. It ends with preserving their purity in the following packaging options, which are free of lubricants and heavy metals. Bulk Pack: Packed in a polyethylene bag that is sealed to perfection
Catalog No. NC1971806 Supplier Cole-Parmer Supplier No. 2450223
Not your everyday graduated pipette tips. It starts with a touch-free manufacturing facility and stringent quality control process to make these clear, plastic tips certified free of any affecting biological elements, such as human DNA, DNase and RNase and Endotoxin, or Pyrogen and PCR inhibitors. They also are noncytotoxic. It’s about their innovative design, too. The profile and dimensions of the tips are engineered to accurately measure and transfer a volume of liquid from one container to another, as well as adapt to a wide-range of pipettes. It ends with preserving their purity in the following packaging options, which are free of lubricants and heavy metals. Bulk Pack: Packed in a polyethylene bag that is sealed to perfection