Learn More
Chemglass Life Sciences Pipette Tip for Research Pipettors - 200 μL Standard Barrier, Sterile, Bulk (4,800/Case)
SDP
Catalog No. 502733214
Encompass_Preferred
Standard 200 μL sterile barrier pipette tips designed for accurate, low-retention liquid handling with most research-grade pipettors. Gamma-irradiated sterility and a hydrophobic aerosol barrier reduce contamination risk, making these tips suitable for molecular biology and general laboratory workflows.
  • 200 μL nominal volume.
  • Made from 100% polypropylene.
  • Gamma-irradiated sterile.
  • Hydrophobic aerosol barrier reduces carry-over and contamination.
  • Certified RNase-free, DNase-free, human DNA-free, and pyrogen-free.
  • Bulk packed, 4,800 tips per case.
  • Low-retention, fine-point design for precise pipetting.
  • Ultra-clear polymer resists common organic solvents.
  • Universal fit for the majority of research-grade pipettors.
Catalog No. 50-273-3214 Supplier Chemglass Life Sciences Supplier No. CLS3499D200
Standard 200 μL sterile barrier pipette tips designed for accurate, low-retention liquid handling with most research-grade pipettors. Gamma-irradiated sterility and a hydrophobic aerosol barrier reduce contamination risk, making these tips suitable for molecular biology and general laboratory workflows.
  • 200 μL nominal volume.
  • Made from 100% polypropylene.
  • Gamma-irradiated sterile.
  • Hydrophobic aerosol barrier reduces carry-over and contamination.
  • Certified RNase-free, DNase-free, human DNA-free, and pyrogen-free.
  • Bulk packed, 4,800 tips per case.
  • Low-retention, fine-point design for precise pipetting.
  • Ultra-clear polymer resists common organic solvents.
  • Universal fit for the majority of research-grade pipettors.