Learn More
Chemglass Life Sciences Chemglass Pipette Tip 300 μL, Standard Barrier, Sterile, 960/Case
SDP
Catalog No. 502733220
Encompass_Preferred
These sterile aerosol-barrier pipette tips are designed for precise molecular biology liquid handling. Made from ultra-clear, premium polypropylene and gamma-irradiated for sterility, they provide low-retention performance and an effective hydrophobic filter to prevent contamination.
  • Sterile, gamma-irradiated for aseptic use.
  • Certified RNase-free, DNase-free, human DNA-free, and pyrogen-free.
  • Low-retention, fine-point design for precise pipetting.
  • Hydrophobic aerosol filter reduces carry-over and contamination.
  • Universal fit for most single and multichannel pipettors.
  • Graduated tips for quick volume checks.
  • Package quantity: 960 per case.
Catalog No. 50-273-3220 Supplier Chemglass Life Sciences Supplier No. CLS3499C300
Add to Cart
Add to Cart
These sterile aerosol-barrier pipette tips are designed for precise molecular biology liquid handling. Made from ultra-clear, premium polypropylene and gamma-irradiated for sterility, they provide low-retention performance and an effective hydrophobic filter to prevent contamination.
  • Sterile, gamma-irradiated for aseptic use.
  • Certified RNase-free, DNase-free, human DNA-free, and pyrogen-free.
  • Low-retention, fine-point design for precise pipetting.
  • Hydrophobic aerosol filter reduces carry-over and contamination.
  • Universal fit for most single and multichannel pipettors.
  • Graduated tips for quick volume checks.
  • Package quantity: 960 per case.