Learn More
Chemglass Life Sciences Chemglass Pipette Tip, Large Volume, 5 mL, Racked, Sterile or Non-sterile (AccuPet compatible)
SDP
Catalog No. NC2952703
Encompass_Preferred
Chemglass Life Sciences large-volume 5 mL pipette tips are supplied in racked format for organized handling and efficient loading. Offered in sterile and non-sterile options, these tips are intended for use with large-volume pipettors to support accurate, high-volume liquid transfers.
  • Racked packaging for organized storage and convenient loading
  • Available sterile or non-sterile to suit lab protocols
  • Designed for accurate 5 mL liquid transfers
  • Compatible with large-volume pipettors and automated dispensing systems
  • Disposable single-use tips for routine laboratory workflows
Catalog No. NC2952703 Supplier Chemglass Life Sciences Supplier No. CLS3499T06
Chemglass Life Sciences large-volume 5 mL pipette tips are supplied in racked format for organized handling and efficient loading. Offered in sterile and non-sterile options, these tips are intended for use with large-volume pipettors to support accurate, high-volume liquid transfers.
  • Racked packaging for organized storage and convenient loading
  • Available sterile or non-sterile to suit lab protocols
  • Designed for accurate 5 mL liquid transfers
  • Compatible with large-volume pipettors and automated dispensing systems
  • Disposable single-use tips for routine laboratory workflows