Learn More
Heathrow Scientific Pipette Tip Disposal Box, Clear
Description
- The 3/8 in. (10 mm) thick acrylic provides protection from Beta radiation
- The large, hinged lid allows easy disposal of contents
- Use in conjunction with small biohazard disposal bags, if hazardous fluids are not acrylic friendly
Specifications
Specifications
| Product Type | Disposal Container |
| Dimensions (L x W x H) | 6.1 x 6.7 x 8.1 in. (15.4 x 17.1 x 20.6 cm) |