Description
The patented Thermo Scientific™ Pipette Tip Disposal Bucket is a functional tool for your lab bench designed specifically to collect used pipette tips during your experiments. The colorful, graffiti themed shield prevents the overshooting of a pipette tip during ejection and guides the tip into the receptacle below. Set up your lab station for maximum efficiency with the ART™ Tip Disposal Bucket for discarded pipette tips.
Specifications
Specifications
| Autoclavable | Not Autoclavable |
| Material | HDPE |
| Height (English) | ˜8.5 inches with shield inserted |
| Diameter (English) Outer | ˜4 inches |