Learn More
Electron Microscopy Sciences Pipette Tip Universal Yellow, 5-200 mcL
SDP
Catalog No. 501899926
Encompass_Preferred
Pipette tips are low binding to ensure accuracy and precision. The low binding surface is the result of a manufacturing process that binds a perfluorooctyl polymer to polypropylene resin–resulting in an exposed trifluoromethyl surface. This surface reduces the surface energy of liquid and repels any substance so sample retention and binding are minimized.
Not detectable/Non-reactive.
Extracted only by a fluorinated solvent.
Autoclaving will not compromise performance.
Available in convenient hinged racked package.
Tips and racks are fully autoclavable. Yellow tips are non-graduated.
Range: 5-200 µl
Catalog No. 50-189-9926 Supplier Electron Microscopy Sciences Supplier No. 7244605
Pipette tips are low binding to ensure accuracy and precision. The low binding surface is the result of a manufacturing process that binds a perfluorooctyl polymer to polypropylene resin–resulting in an exposed trifluoromethyl surface. This surface reduces the surface energy of liquid and repels any substance so sample retention and binding are minimized.
Not detectable/Non-reactive.
Extracted only by a fluorinated solvent.
Autoclaving will not compromise performance.
Available in convenient hinged racked package.
Tips and racks are fully autoclavable. Yellow tips are non-graduated.
Range: 5-200 µl