Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BrandTech™ BRAND™ Pipette Tips, 0.5 to 20 μL

Catalog No. 13878864 Shop All BrandTech Scientific Products
Change view
Click to view available options
Volume (Metric):
0.5 to 20 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
13-878-864 0.5 to 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 13-878-864 Supplier BrandTech™ BRAND™ Supplier No. 732124
Add to Cart
Add to Cart

Manufactured from high-purity, oleamide and diHEMDA-free polypropylene. Pipet Tips, 0.5 to 20μL allow pipetting in microtubes and microplates without touching the walls. Gradations marked at 2μL and 10μL provide quick volume assessment.

  • Nonsterile , high-purity polypropylene is free from oleamide and diHEMDA.
  • Gradations marked at 2 μL and 10 μL provide quick volume assessment.
  • Slender design allows pipetting in microtubes and microplates without touching the walls.
  • Available in either standard or ultra low retention types.
  • Available either in bulk packages or in convenient rack-and-stack formats.

Ultra low retention tips

  • Produced through special, patented physicochemical process.
  • Homogeneous, defect-free surface thus produced has low surface tension.
  • Reduce sample loss and provide substantially higher reproducibility when working with critical media.
  • For biological samples that contain detergents such as Triton™ X-100, SDS, Tween, etc.
  • High chemical resistance.
  • Tips can be autoclaved at 121°C (2 bar) without damaging material properties.
TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 0.5 to 20 μL
Filtered Non-filtered
Sterility Sterile
Color Colorless
Rack Color Gray
Autoclavable Autoclavable
Length (English) 1.8 in.
Length (Metric) 46 mm
Clean Claims DNase, RNase, ATP, and Endotoxin Free
For Use With (Application) Reverse pipetting, plate washing, working with multichannel pipettes
Format Refill
Tip Style Narrow
Packaging TipRack
Quantity 960/Pk.
Product Type Pipette Tips
Disposable Limited-use
For Use With (Equipment) Universal Fit Ultramicro
Graduated Yes
Material Polypropylene
Fit for Purpose PCR Ready
Graduations 2, 10 μL
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.