Learn More
Mascon Medical Mascon Medical™ Rainin Compatible Pipette Tips, Filtered, Sterile, 1000ul, 96EA/Rack, 10 Racks/Box, 5 Boxes/Case
SDP
Supplier:  Mascon Medical FSRAI1000
Encompass_Preferred
Mascon Medical™ offers an extensive line of compatible robotic tips for Hamilton, Eppendorf™, Rainin, and Beckman automation systems. These high-precision tips are manufactured to guarantee a perfect fit and extremely accurate dispensing behavior. Available in non-filtered and filtered. Nonpyrogenic. Conductive Polypropylene tips are available in 96-well nested tip rack.Features:•96 Tip/Nested Tip Rack Format•Meets ISO13485 quality management system standard•Nonpyrogenic•Low-retention•Rainin CompatibleSpecifications:•Volume: 1000µL•Qty. / Case: 4800•Size: 101.7mm•Tip Format: 96 Tips/Rack•Material: Polypropylene•Temperature Range: -40℃~100℃•Color: Blue•Filter: Yes•Sterile: Yes•Packaging: Nested Tip Rack, 96EA/Rack, 10 Racks/Box, 5 Boxes /Case•Packaging Format: 50 Racks/Case
Catalog No. NC2458318