Learn More
MTC Bio 10mL Pipette Tips for Halo™ Pipettors, 100/pk
Catalog No. NC2017345
Encompass_Preferred
These new tips are designed to fit MTC’s exclusive 300µL single channel pipettes.  Many researchers  have recognized the need for an accurate pipette for volumes above 200µL, which is a range that is not well served by traditional 1mL pipettes. The MTC Bio 300µL pipette and pipette tips close the gap and permit accurate and convenient pipetting, especially in the 200-300µL range. These tips also fit most popular 300µL multi-channel pipettes and are supplied pre-sterilized in 96 position racks or refill wafers.
Catalog No. NC2017345 Supplier MTC Bio Supplier No. H6510
These new tips are designed to fit MTC’s exclusive 300µL single channel pipettes.  Many researchers  have recognized the need for an accurate pipette for volumes above 200µL, which is a range that is not well served by traditional 1mL pipettes. The MTC Bio 300µL pipette and pipette tips close the gap and permit accurate and convenient pipetting, especially in the 200-300µL range. These tips also fit most popular 300µL multi-channel pipettes and are supplied pre-sterilized in 96 position racks or refill wafers.