Learn More
Alkali Scientific Pipette Tips, 10mL, 150/cs
SDP
Numéro de catalogue. 502389444
Encompass_Preferred
Macro 10mL tips are specially designed for an ideal and comfortable fit on our Macro size pipettes. Supplied in bags, these tips are made of ultra-clear polypropylene and are guaranteed to create the most ideal seal on ProPette™ Pipettors.
Numéro de catalogue. 50-238-9444 Fournisseur Alkali Scientific Code fournisseur P9310
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
Macro 10mL tips are specially designed for an ideal and comfortable fit on our Macro size pipettes. Supplied in bags, these tips are made of ultra-clear polypropylene and are guaranteed to create the most ideal seal on ProPette™ Pipettors.
missing translation for 'productTitle'
missing translation for 'pleaseSelectIssue'
missing translation for 'byClickingSubmit' missing translation for 'privacyPolicy'.