Learn More
Chemglass Life Sciences Chemglass pipette tips, 10 μL extra-long, non-sterile refill, 960/pack
SDP
Catalog No. 502733206
Encompass_Preferred
Extra-long 10 μL pipette tips supplied as refill wafers for high-precision liquid handling and access to narrow vessels.
  • Extra-long fine-point geometry reduces sample adhesion and improves access to narrow tubes.
  • Ultra-clear polymer resists organic solvents for clear, error-free pipetting.
  • Low-retention surface helps improve accuracy and sample recovery.
  • Non-sterile refill format: 960 tips per pack, 9,600 per case.
  • Hydrophobic aerosol barrier design reduces carry-over and contamination during pipetting.
Catalog No. 50-273-3206 Supplier Chemglass Life Sciences Supplier No. CLS3499G010
Extra-long 10 μL pipette tips supplied as refill wafers for high-precision liquid handling and access to narrow vessels.
  • Extra-long fine-point geometry reduces sample adhesion and improves access to narrow tubes.
  • Ultra-clear polymer resists organic solvents for clear, error-free pipetting.
  • Low-retention surface helps improve accuracy and sample recovery.
  • Non-sterile refill format: 960 tips per pack, 9,600 per case.
  • Hydrophobic aerosol barrier design reduces carry-over and contamination during pipetting.