Learn More
Chemglass Life Sciences Pipette Tips, 1,250 μL Refill Nest for Large-Volume Pipettors
SDP
Catalog No. 502733203
Encompass_Preferred
Clear, non-sterile pipette tip refills with a 1,250 μL nominal capacity sold in nested packs for bulk laboratory use. Designed for general large-volume liquid handling and to fit most single- and multichannel pipettors.
  • 1,250 μL nominal capacity.
  • Non-sterile.
  • Sold in nested packs: 960 pieces per pack, 4,800 pieces per case.
  • Universal fit for most single- and multichannel pipettors.
  • Intended for large-volume liquid handling and refill applications.
Catalog No. 50-273-3203 Supplier Chemglass Life Sciences Supplier No. CLS3499G125
Add to Cart
Add to Cart
Clear, non-sterile pipette tip refills with a 1,250 μL nominal capacity sold in nested packs for bulk laboratory use. Designed for general large-volume liquid handling and to fit most single- and multichannel pipettors.
  • 1,250 μL nominal capacity.
  • Non-sterile.
  • Sold in nested packs: 960 pieces per pack, 4,800 pieces per case.
  • Universal fit for most single- and multichannel pipettors.
  • Intended for large-volume liquid handling and refill applications.