Learn More
Med Vet International Pipette Tips, 1ml, J0857A, 500Pack
Catalog No. 5000581948
Encompass_Preferred
Pipettors are widely used in laboratories. They are an accurate method of transferring solutions or reagents. They also reduce exposure to samples or laboratory chemicals.Consisting of two parts, a reusable pipettor draws up fluid through the disposable tip which actually holds the fluid in a preset, precise volume. The fluid never reaches or touches the actual pipettor. The disposable pipettes cost only pennies and are discarded after each use. Pipettors are available in either fixed or adjustable volume.Pipette Tip.1.0ml Capacity Pipette Tip. 500/pk.
Catalog No. 50-005-81948 Supplier Med Vet International Supplier No. JOR857A
Pipettors are widely used in laboratories. They are an accurate method of transferring solutions or reagents. They also reduce exposure to samples or laboratory chemicals.Consisting of two parts, a reusable pipettor draws up fluid through the disposable tip which actually holds the fluid in a preset, precise volume. The fluid never reaches or touches the actual pipettor. The disposable pipettes cost only pennies and are discarded after each use. Pipettors are available in either fixed or adjustable volume.Pipette Tip.1.0ml Capacity Pipette Tip. 500/pk.