Learn More
Mascon Medical Mascon Medical™ Rainin Compatible Pipette Tips, 200ul, Sterile, Filtered, 96EA/Rack, 10 Racks/Box, 5 Boxes/Case
SDP
Catalog No. NC2458325
Encompass_Preferred
Mascon Medical™ offers an extensive line of compatible robotic tips for Hamilton, Eppendorf™, Rainin, and Beckman automation systems. These high-precision tips are manufactured to guarantee a perfect fit and extremely accurate dispensing behavior. Available in non-filtered and filtered. Nonpyrogenic. Conductive Polypropylene tips are available in 96-well nested tip rack.Features:•96 Tip/Nested Tip Rack Format•Meets ISO13485 quality management system standard•Compatible with Rainin•Nonpyrogenic•Low-retentionSpecifications:•Volume: 200µL•Qty. / Case: 4800•Size: 49.9mm•Tip Format: 96 Tips/Rack•Material: Polypropylene•Temperature Range: -40℃~100℃•Color: Green•Filter: Yes•Sterile: Yes•Packaging: Nested Tip Rack, 10 Racks/Pack, 5 Packs /Carton•Packaging Format: 50 Racks/Carton
Catalog No. NC2458325 Supplier Mascon Medical Supplier No. FSRAI200S
Mascon Medical™ offers an extensive line of compatible robotic tips for Hamilton, Eppendorf™, Rainin, and Beckman automation systems. These high-precision tips are manufactured to guarantee a perfect fit and extremely accurate dispensing behavior. Available in non-filtered and filtered. Nonpyrogenic. Conductive Polypropylene tips are available in 96-well nested tip rack.Features:•96 Tip/Nested Tip Rack Format•Meets ISO13485 quality management system standard•Compatible with Rainin•Nonpyrogenic•Low-retentionSpecifications:•Volume: 200µL•Qty. / Case: 4800•Size: 49.9mm•Tip Format: 96 Tips/Rack•Material: Polypropylene•Temperature Range: -40℃~100℃•Color: Green•Filter: Yes•Sterile: Yes•Packaging: Nested Tip Rack, 10 Racks/Pack, 5 Packs /Carton•Packaging Format: 50 Racks/Carton