Learn More
Chemglass Life Sciences Chemglass Pipette Tips Refill - 200 μL, Clear, Non-Sterile (960/Pack)
SDP
Catalog No. 502733202
Encompass_Preferred
Refill pack of clear 200 μL pipette tips designed for general laboratory pipetting and bulk restocking of tip boxes or racks.
  • 200 μL nominal volume.
  • 960 pieces per pack; 9,600 pieces per case.
  • Non-sterile bulk refills.
  • Ultra-clear polypropylene construction for chemical resistance and visibility.
  • Low-retention, fine-point geometry reduces sample adhesion.
  • Universal fit for most single- and multichannel pipettors.
Catalog No. 50-273-3202 Supplier Chemglass Life Sciences Supplier No. CLS3499G200
Add to Cart
Add to Cart
Refill pack of clear 200 μL pipette tips designed for general laboratory pipetting and bulk restocking of tip boxes or racks.
  • 200 μL nominal volume.
  • 960 pieces per pack; 9,600 pieces per case.
  • Non-sterile bulk refills.
  • Ultra-clear polypropylene construction for chemical resistance and visibility.
  • Low-retention, fine-point geometry reduces sample adhesion.
  • Universal fit for most single- and multichannel pipettors.