Learn More
Mascon Medical Pipette Tips LTS, Sterile, Filtered,8 racks/pack #30374969, 300L, 40 Rack/Case
SDP
Catalog No. NC2180267
Encompass_Preferred
Mascon Medical™ offers an extensive line of compatible robotic tips for Hamilton, Eppendorf™, Rainin and Beckman automation systems. These high precision tips are manufactured to guarantee a perfect fit and extremely accurate dispensing behavior. Available in non-filtered and filtered. Nonpyrogenic. Conductive Polypropylene tips are available in 96-well nested tip rack.Features: 96 Tip/Nested Tip Rack Format Nonpyrogenic Sterile Filtered Specifications: Volume: 300L Qty. / Case: 3840 Tip Shape: Round Tip Format: 96 Tips/Rack Material: Polypropylene Color: Yellow Filter: Yes Sterile: Yes Packaging: Nested Tip Rack, 8 Racks/Pack, 5 Packs /Carton Packaging Format: 40 Racks/Carton
This item is not returnable. View return policy
Catalog No. NC2180267 Supplier Mascon Medical Supplier No. MMPTRAININ300
Mascon Medical™ offers an extensive line of compatible robotic tips for Hamilton, Eppendorf™, Rainin and Beckman automation systems. These high precision tips are manufactured to guarantee a perfect fit and extremely accurate dispensing behavior. Available in non-filtered and filtered. Nonpyrogenic. Conductive Polypropylene tips are available in 96-well nested tip rack.Features: 96 Tip/Nested Tip Rack Format Nonpyrogenic Sterile Filtered Specifications: Volume: 300L Qty. / Case: 3840 Tip Shape: Round Tip Format: 96 Tips/Rack Material: Polypropylene Color: Yellow Filter: Yes Sterile: Yes Packaging: Nested Tip Rack, 8 Racks/Pack, 5 Packs /Carton Packaging Format: 40 Racks/Carton