Learn More
Agilent BioTek Pipette Tips for KC4 Automated Pipetting System
Shop All Agilent Technologies Products

Description
Order Info
96 tips per rack, 10 racks per case
Specifications
Specifications
| Volume (Metric) | 250 μL |
| Sterility | Non-sterile |
| Format | Rack |
| For Use With (Equipment) | BioTek Precision, BioTek Precision XS |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.