Learn More
Thomas Scientific Transfer Pipet, 7.0mL, Large Bulb, Graduated to 3mL, 155mm, Bulb Draw - 3.2mL
SDP
Catalog No. 501460815
Encompass
Material: Low Density Polyethylene (LDPE) Packaging: Globe Scientific's non-sterile transfer pipets are packaged in a unique dispenser box. This convenient box is small and compact and ideal for bench-top use. The hinged front panel opens for quick access to the pipets and can be closed when not in use to keep the contents clean. Each dispenser box contains 500 pipets.
Catalog No. 50-146-0815 Supplier Thomas Scientific Supplier No. 1193U93
Material: Low Density Polyethylene (LDPE) Packaging: Globe Scientific's non-sterile transfer pipets are packaged in a unique dispenser box. This convenient box is small and compact and ideal for bench-top use. The hinged front panel opens for quick access to the pipets and can be closed when not in use to keep the contents clean. Each dispenser box contains 500 pipets.