Learn More
Med Vet International McKesson Transfer Pipette, 5 mL, 0.5 to 1 mL Graduation Increments, NonSterile, 558, 500/Box
Catalog No. 503043096
Encompass_Preferred
McKesson Transfer Pipette 5 mL 0.5 to 1 mL Graduation Increments NonSterile McKesson transfer pipettes are one-piece and non-sterile: They are produced from a low density polyethylene which will not shatter or break5 mL volume with 3.4 draw volume Nonsterile
Catalog No. 50-304-3096 Supplier Med Vet International Supplier No. 911824BX
McKesson Transfer Pipette 5 mL 0.5 to 1 mL Graduation Increments NonSterile McKesson transfer pipettes are one-piece and non-sterile: They are produced from a low density polyethylene which will not shatter or break5 mL volume with 3.4 draw volume Nonsterile