Learn More
Med Vet International McKesson Transfer Pipette, 7.5 mL, 0.5 to 3 mL Graduation Increments, NonSterile, 559, 5000/Case
Catalog No. 503043757
Encompass_Preferred
McKesson Transfer Pipette 7.5 mL 0.5 to 3 mL Graduation Increments NonSterile Liquids do not stick to these McKesson 7.5 mL non-sterile transfer pipettes The pipettes can be can easily access large bottles or long-necked containers thanks to their long stems The 3mL draw volume capacity allows you to suck large volumes of thick liquid at one time These McKesson polyethylene transfer pipettes will not react with biological fluids and most acids Their flexible stems are durable and do not shatter or break easily
Catalog No. 50-304-3757 Supplier Med Vet International Supplier No. 911826CS
McKesson Transfer Pipette 7.5 mL 0.5 to 3 mL Graduation Increments NonSterile Liquids do not stick to these McKesson 7.5 mL non-sterile transfer pipettes The pipettes can be can easily access large bottles or long-necked containers thanks to their long stems The 3mL draw volume capacity allows you to suck large volumes of thick liquid at one time These McKesson polyethylene transfer pipettes will not react with biological fluids and most acids Their flexible stems are durable and do not shatter or break easily