Learn More
Med Vet International McKesson Transfer Pipette, 7.5 mL, 0.5 to 3 mL Graduation Increments, NonSterile, 559, 500/Box
Catalog No. 503043756
Encompass_Preferred
McKesson Transfer Pipette 7.5 mL 0.5 to 3 mL Graduation Increments NonSterile Liquids do not stick to these McKesson 7.5 mL non-sterile transfer pipettes The pipettes can be can easily access large bottles or long-necked containers thanks to their long stems The 3mL draw volume capacity allows you to suck large volumes of thick liquid at one time These McKesson polyethylene transfer pipettes will not react with biological fluids and most acids Their flexible stems are durable and do not shatter or break easily
Catalog No. 50-304-3756 Supplier Med Vet International Supplier No. 911826BX
McKesson Transfer Pipette 7.5 mL 0.5 to 3 mL Graduation Increments NonSterile Liquids do not stick to these McKesson 7.5 mL non-sterile transfer pipettes The pipettes can be can easily access large bottles or long-necked containers thanks to their long stems The 3mL draw volume capacity allows you to suck large volumes of thick liquid at one time These McKesson polyethylene transfer pipettes will not react with biological fluids and most acids Their flexible stems are durable and do not shatter or break easily